Find methods for your needs
AB001809: Improved Determinations of Residual Anions and Organic Acids to Improve Printed Circuit Board Cleanliness - A
Description
Organic acid and inorganic residues from solder flux and other manufacturing processes can cause soft and hard failures in the printed circuit board (PCB), as previously discussed in Thermo Scientific Application Note 1163. This Application Brief demonstrates methods to improve analyte resolution. This record shows the effect of reducing column temperature on analyte resolution.Market: | Other |
Keywords: | Formate, Malate, Organic acids, IonPac AS11-HC column, fluoride, Succinate, Nitrate, Phthalate, RFIC, Nitrite, Chloride, phosphate, Acetate, Sulfate, adipate, methanesulfonate, Bromide, AN1163, IPC-TM-650, AB 001809, AB001809 |
Matrix: | DI water |
Author: | Terri Christison and Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.