Find methods for your needs
AB001809: Improved Determinations of Residual Anions and Organic Acids to Improve Printed Circuit Board Cleanliness -B Be the first to rate this application
Description
Organic acid and inorganic residues from solder flux and other manufacturing processes can cause soft and hard failures in the printed circuit board (PCB), as previously discussed in Thermo Scientific Application Note 1163. This Application Brief demonstrates methods to improve analyte resolution. This record shows the effect of adding methanol to the eluent to improve analyte resolution.
Market: Other
Keywords: Formate, Malate, Methanol, Organic acids, IonPac AS11-HC column, fluoride, Succinate, Nitrate, Phthalate, RFIC, Nitrite, Chloride, phosphate, Acetate, Sulfate, adipate, methanesulfonate, Bromide, AN1163, IPC-TM-650, AB 001809, AB001809
Matrix: DI water
Author: Terri Christison and Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Status: Proof of Concept
Uploaded on 2/9/2023.
For Research Use Only. Not for use in diagnostic procedures.