Find methods for your needs
Orthogonal method for degraded impurities analysis of Teriparatide by IEX-HPLC-FLD
Description
Ion-exchange high-performance liquid chromatography (IEX-HPLC) analysis were performed to quantitate degraded impurities of Teriparatide with a fluorescence detector. The analysis was performed on a Thermo Scientific MAbPac SCX-10 RS HPLC column. The separation results were satisfactory.| Market: | BioPharma |
| Keywords: | HPLC, FLD detection, Teriparatide |
| Matrix: | drug dosage |
| Author: | Krishna Rao Dara |
| Affiliation: | Thermo Fisher Scientific, CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.