Find methods for your needs
AN72714: HPAE-PAD Analysis of Galactosyloligosaccharide-Containing Samples using Dual Eluent Generation Cartridge Mode
Description
Our study demonstrated GOS separation using Dual EGC mode and a Dionex CarboPac PA200 1 mm column. The results reported here update an earlier application note that used a GOS sample containing maltodextrin and incorrectly reported maltodextrin peaks as GOS peaks.| Market: | Food and Beverage |
| Keywords: | GOS, oligosaccharide, HPAE-PAD, prebiotics, Dionex ICS-6000, Dionex EGC 400 KOH, Dionex EGC 400 MSA, TN71, Dionex CarboPac PA200 column (1 mm i.d.), TN 71, Dual EGC Mode, TN 20, TN 70671, TN20, TN70671, AN 72714 |
| Matrix: | Food |
| Author: | Manali Aggrawal, Sachin Patil, and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.