Find methods for your needs
Related Impurities analysis of Dutasteride and Tamsulosin hydrochloride using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the related impurities analysis of dutasteride and tamsulosin hydrochloride. The analysis was performed on a Thermo Scientific Acclaim 120 C18 HPLC column. The separation results and peak asymmetry were satisfactory| Market: | Pharma |
| Keywords: | Dutasteride, HPLC, UV detection, Tamsulosin |
| Matrix: | N/A |
| Author: | Krishna Rao Dara |
| Affiliation: | thermo Scientific, CoE Lab, Ahmedabad, India |