Find methods for your needs
Chromatographic method for size base impurity analysis of Filgrastim
Description
The Thermo Scientific Dionex ultimate 3000 Bio LC system is Applied for the Aggregate impurities analysis of Filgrastim. The analysis was performed on Mabpac SEC-1 using the IP monograph method. IP has revised the resolution criteira from 2.0 to 3.0. The critical resolution of Aggregate and monomer was more than 3.0 .| Market: | BioPharma |
| Matrix: | NA |
| Author: | Mazin Adenwala |
| Affiliation: | Thermo Fisher Scientific, CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.