Find methods for your needs
AN001054: Determination of Organochlorine Pesticides (OCPs) in Soils using the EXTREVA ASE Accelerated Solvent Extractor and GC-ECD
Description
This application described a method for the determination of organochlorine pesticides (OCPs) in soil matrices using the Thermo Scientific EXTREVA ASE Accelerated Solvent Extractor with Gas Chromatograph and electron capture detector (ECD). Good recovery and reproducibility were observed for all analytes. Carryover between consecutive runs were minimal. And thermal degradation was low under the extraction conditions. By combining two sample preparation instruments into one, the EXTREVA ASE performs both extraction and evaporation for organic compounds in one operation.| Market: | Environmental |
| Keywords: | environmental, Pressurized Fluid Extraction, TRACE 1310 Gas Chromatograph, automated sample preparation, electron capture detector (ECD), U.S. EPA Method 8081B, OCPs |
| Matrix: | Soil |
| Author: | Rahmat Ullah, Fabrizio Galbiati, Anzi Wang, Changling Qiu, Mingfang Wang, Yan Liu |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA and Shanghai, PRC |
For Research Use Only. Not for use in diagnostic procedures.