![](/Content/Images/icons/rate.png?v=2511129)
![](/Content/Images/icons/ask_expert.png?v=2511129)
![](/Content/Images/icons/request.png?v=2511129)
![](/Content/Images/icons/favorites.png?v=2511129)
![](/Content/Images/icons/create_parts_list.png?v=2511129)
Parts List
![](/Content/Images/icons/send_email.png)
Friend
![](/Content/Images/icons/print.png?v=2511129)
Find methods for your needs
![](/Content/Images/icons/search_help.png?v=2511129)
AN001106: Determination of Polycyclic Aromatic Hydrocarbons in Soils using the EXTREVA ASE Accelerated Solvent Extractor and GC-MS
![determination pahs](/Image/AppImage/4760/20221107111336/determination-pahs?imageNameOnDisk=d0963d0c-40bd-4a59-9d17-36cb69929d2f-02eaf8cf-66ed-4edd-9b77-ba04d99f29cf.png)
Description
This application note described a method for the determination of polycyclic aromatic hydrocarbons (PAHs) in soil matrices using the Thermo Scientific EXTREVA ASE Accelerated Solvent Extractor with Gas Chromatograph and Mass Spectrometer. Good recovery and reproducibility were observed for all analytes. Carryover between consecutive runs was minimal. By combining two sample preparation instruments into one, the EXTREVA ASE system performs both extraction and evaporation for organic compounds in one seamless operation.Market: | Environmental |
Keywords: | PAHs, environmental, soil, Pressurized Fluid Extraction, Sample preparation, ISQ 7000 Single Quadrupole GC-MS, U.S. EPA Method 8270 |
Matrix: | Soil, solid |
Author: | Changling Qiu, Anzi Wang, Rahmat Ullah, Yan Liu, Mingfang Wang, Dustin Lu |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA and Shanghai, PRC |
For Research Use Only. Not for use in diagnostic procedures.