Find methods for your needs
AN001419: Determination of Polychlorinated Biphenyls (PCBs) in Soils using a New Fully Automated Parallel Extraction and Evaporation System and GC-MS
Description
This application note described a method for the determination of PCBs in soil matrices using the Thermo Scientific EXTREVA ASE Accelerated Solvent Extractor with Gas Chromatograph and Mass Spectrometer. Good recovery and reproducibility were observed for all analytes. Carryover between consecutive runs was minimal. By combining two sample preparation instruments into one, the EXTREVA ASE system performs both extraction and evaporation for organic compounds in one seamless operation.| Market: | Environmental |
| Keywords: | environmental, soil, Pressurized Fluid Extraction, PCBs, Sample preparation, ISQ 7000 Single Quadrupole GC-MS, U.S. EPA Method 8082A, AN 001419 |
| Matrix: | Soil, solid |
| Author: | Mingfang Wang, Rahmat Ullah, Fabrizio Galbiati, Xiangying Meng, Duanjun Lu, Panfeng Jiang, Changling Qiu |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.