Find methods for your needs
AN001166: Determination of Cationic Polar Pesticides in Oat Cereals by Ion Chromatography and Electrospray Ionization Mass Spectrometry
Description
Pesticide contamination in food as a potential health risk is a growing public concern, resulting in increased interest and attention by health researchers and regulatory agencies. The Thermo Scientific™ Dionex™ IonPac™CS21-Fast-4µm column selected for this method, was optimized to fully resolve paraquat from diquat. In this application, a targeted screening method is demonstrated for cationic polar pesticides in oat cereals using the IonPac CS21-Fast-4µm column, the Dionex ICS-6000 HPIC system combined with the Thermo Scientific™ ISQ EC™ single quadrupole mass spectrometer.| Market: | Food and Beverage |
| Keywords: | Diquat, Mepiquat, Quaternary amines, RFIC, IC-MS, Paraquat, ISQ EC mass spectrometer, chlormequat, cationic pesticides, TN73990, Reagent-Free IC (IC), cationic polar pesticides, CR-CTC III trap column, EURL-FV, SANTE, AN 001166, IonPac CS21-Fast-4µm column, AN000607 |
| Matrix: | Extract of oatmeal cereal |
| Author: | Terri Christison, John E. Madden, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.