Find methods for your needs
AN001509: Anion Exchange-Based Improved Method for Nucleotide Sugar Determination
Description
This application note describes a method for nucleotide sugar determination using anion-exchange chromatography. This method uses a Thermo Scientific™ Dionex™ CarboPac™ PA1 column and manually prepared eluents for separation. Using a this column, a 34 min method that resolved seven analytes was designed.| Market: | BioPharma |
| Keywords: | CarboPac PA1, HPAE, AN 001509 |
| Matrix: | DI water |
| Author: | Sachin Patil and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA, USA |