Find methods for your needs
Simultaneous analysis of drug substances according to USP assay and impurity methods
Description
This application note describes a comprehensive analysis of a drug substance. Specifically, the analysis requires two different eluent compositions for the determination of drug content and impurities of esomeprazole magnesium according to the United States Pharmacopeia (USP) monograph. With conventional (U)HPLC systems, this would either require two different systems or sequential analyses.| Market: | Pharma |
| Keywords: | Esomeprazole, HPLC, Omeprazole, USP, Impurities, Assay, productivity, Vanquish Duo for Dual LC, parallel analysis, throughput, pharmacopeial monograph |
| Matrix: | Acetonitrile, phosphate buffer |
| Author: | Sylvia Grosse*, Alessandra Mantani*, Mauro De Pra*, Bhushan Pandit**, Frank Steiner* |
| Affiliation: | *Thermo Fisher Scientific, Germering, Germany; **Thermo Fisher Scientific, Mumbai, India |
For Research Use Only. Not for use in diagnostic procedures.