Find methods for your needs
AB001056: Citric Acid Determination in Mixed Beer Beverages
Description
This record presents a fast isocratic method to determine citric acid in carbonated beverages with a carbonate-based eluent and electrolytically regenerated suppression. This application can be run on any Thermo Scientific™ Dionex™ IC system equipped with suppressed conductivity detection. The carbonate eluent (see Method) can be generated electrolytically using Thermo Scientific™ Dionex™ RFIC™ technology.| Market: | Food and Beverage |
| Keywords: | isocratic, Suppressed Conductivity, carbonate eluent, Dionex AERS 500 Carbonate Suppressor, Aquion, 2 mm columns, continuous electrolytic regeneration, Dionex IonPac AS29-Fast-4µm column |
| Matrix: | Carbonated beverage |
| Author: | Beate Appel; Christian Weller; Detlef Jensen |
| Affiliation: | Thermo Fisher Scientific, Dreieich Germany |