Find methods for your needs
Analysis of free polyethylene glycol in recombinant protein pharmaceutical preparations by CAD (电雾式检测器用于聚乙二醇重组蛋白药物制剂中游离聚乙二醇测定)
Description
Because PEG has no uv absorption, it is important to select a universal detector with high sensitivity and high repeatability for quality control of free PEG in polypeptide drugs. CAD has been successfully detected in a variety of different molecular weight PEG. In this paper, 20000 molecular weight PEG detection was tried for the first time, and the performance of ELSD detection was preliminarily compared.Market: | BioPharma |
Keywords: | CAD, PEG20000, polypeptide drugs |
Matrix: | recombinant protein pharmaceutical preparations |
Author: | ruiping.chai; jianming.cao; yan.jin |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.