Find methods for your needs
AN22017: Determination of lithium salt and its additives in electrolytes by IC (AN22017: 离子色谱法测定电解液中锂盐及锂盐添加剂的含量)
Description
In this application, a method for the detection the concentrations of lithium salt and its additives in electrolytes of lithium batteries has been developed. This method is using the Thermo Scientific Dionex Aquion IC system. For the separation, a Thermo Scientific Dionex IonPac AS22 column with carbonate eluent isocratic elution was used. An analysis can be completed within 23 minutes. This method is suitable for routine analysis in the lithium battery industry.| Market: | Energy |
| Keywords: | Aquion, Electrolyte, 电解液, Lithium batteries, Lithium salt, Lithium salt additive, Methyl ethyl carbonate, 锂电池, 锂盐, 锂盐添加剂, Aquion 离子色谱仪, 碳酸甲乙酯 |
| Matrix: | lithium battery electrolyte, 锂电池电解液 |
| Author: | Guoquan.Wu (吴国权); Hongguo.Zheng (郑洪国); Jie.Chen (陈洁) |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.