Find methods for your needs
AN22019: Rapid determination of lithium salt and its additives in electrolytes by HPIC (AN22019: 高压离子色谱快速测定电解液中锂盐及锂盐添加剂的含量)
Description
In this application note, a fast method for the determination of lithium salt and its additives has been developed using a Thermo Scientific Dionex Integrion HPIC system. This method realizes the analysis of lithium salt and its additives in only 9 minutes. The method uses a Thermo Scientific Dionex IonPac AS22-Fast-4µm column. This application is suitable for analyses with high sample throughput in the lithium battery industry.| Market: | Energy |
| Keywords: | Integrion, Dionex IonPac AS22-Fast-4μm, Electrolyte, 电解液, Lithium batteries, Lithium salt, Lithium salt additive, Methyl ethyl carbonate, 锂电池, 锂盐, 锂盐添加剂, 碳酸甲乙酯 |
| Matrix: | lithium battery electrolyte, 锂电池电解液 |
| Author: | Guoquan.Wu (吴国权); Hongguo.Zheng (郑洪国); Jie Chen(陈洁) |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.