Find methods for your needs
Fast analysis of fluorinated nucleic bases using a Thermo Scientific Hypersil GOLD PFP HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of fluorinated nucleic bases. The separation is performed on a Thermo Scientific Hypersil GOLD PFP HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Fluorinated nucleic bases, Fluorocytosine, Fluorouracil, HPLC, Hypersil Gold PFP, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.