Find methods for your needs
Unstoppable analysis of pesticides residues in black tea using triple quadrupole GC-MS
Description
In this study, the suitability of the TSQ 9610 triple quadrupole GC-MS/MS system was assessed for the analysis of more than 200 pesticides in black tea at trace concentrations supplemented with SPE cleanup. The linearity, accuracy, precision, limit of quantitation, and injection reproducibility of 20 selected analytes representative of the different pesticide classes in black tea matrix were demonstrated.| Market: | Food and Beverage; Environmental |
| Keywords: | food, Solid phase extraction, Persistent Organic Pollutants, Current use pesticides, QuEChERS extracton, SANTE/12682/2019 |
| Matrix: | Black tea |
| Author: | Camille Grigsby(1), Carlos Parra(1), Jerry Mueller(1), Katie Banaszewski(1), Giulia Riccardino(2) and Adam Ladak(3) |
| Affiliation: | 1. NOW Foods, Bloomingdale, IL, USA; 2. Thermo Fisher Scientific, Milan, IT; 3. 3Thermo Fisher Scientific, Hemel Hempstead, UK |
For Research Use Only. Not for use in diagnostic procedures.