Find methods for your needs
Analysis of volatile organic compounds in soil using purge and trap coupled to single quadrupole GC-MS
Description
To demonstrate the compliance of the Teledyne Tekmar Atomx XYZ purge and trap (P&T) system for online extraction and analysis of volatile organic compounds (VOCs) in soil with the Thermo Scientific™ ISQ™ 7610 single quadrupole mass spectrometer. Separation of targeted VOCs was achieved using the Thermo Scientific™ TRACE™1610 gas chromatograph (GC). Linearity, detection limit, precision, and accuracy were assessed to evaluate method performance in accordance with Chinese Method HJ 605.| Market: | Environmental |
| Keywords: | volatile organic compounds (VOCs), Chinese Method HJ 605, online extraction |
| Matrix: | Soil |
| Author: | Terry Jeffers (1), Adam Ladak(2), and Amy Nutter(3) |
| Affiliation: | 1. Thermo Fisher Scientific, USA; 2. Thermo Fisher Scientific, UK; 3. Teledyne Tekmar, Mason, OH, USA |
For Research Use Only. Not for use in diagnostic procedures.