Find methods for your needs
AU000610: Determination of Trace Organic Acids and Inorganic Anions in Boric Acid-Treated Power Plant Waters using an Automated Reagent-Free Ion Chromatography System Be the first to rate this application
Description
Ion chromatography (IC) is an indispensable technique for the determination of trace concentrations of ionic impurities found in power plant waters. Dionex AN185 describes an automated RFIC method for the determination of trace anions in borated waters. This work updates AN185 with a Dionex IonPac AS28-Fast-4μm column for improved peak efficiencies and resolution, the IC instrument, and consumables. The calibration ranges, detection limits, precision, and accuracy of this method for determining trace anionic impurities in borated samples are described.
Market: Electronics & Semiconductors
Keywords: trace analysis, Dionex ICS-6000 HPIC system, ADRS 600 Suppressor, Dionex IonPac AS28-Fast-4μm column, AU 000610
Matrix: boric acid-treated power plant waters
Author: Jingli Hu and Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Uploaded on 3/28/2022.
For Research Use Only. Not for use in diagnostic procedures.