Find methods for your needs
A rapid, sensitive, and consolidated method for PCBs and PAHs using GC-MS/MS
Description
Determination of PAHs and PCBs in water using the TSQ 9610 GC-MS/MS. The sample preparation procedure and chromatographic methods were optimized to reduce overall analysis time. Linearity, instrument detection limits (IDLs), robustness and reproducibility were assessed for all targeted compounds for trace analysis of PCBs and PAHs in water matrices.| Market: | Water Analysis; Environmental |
| Keywords: | Liquid-liquid extraction, Polychlorinated biphenyls (PCBs), Persistent Organic Pollutants, NeverVent Advanced Ionization ion source (AEI), Polyaromatics hydrocarbons (PAHs), programable temperature vaporization (PTV) |
| Matrix: | Water |
| Author: | Delphine Thomas¹, Bénédicte Gauriat¹, Aristide Ganci¹, Adam Ladak², and Jean-François Garnier¹ |
| Affiliation: | ¹Thermo Fisher Scientific Customer Solution Center, Les Ulis, France; ²Thermo Fisher Scientific, Hemel Hempstead, UK |
For Research Use Only. Not for use in diagnostic procedures.