Find methods for your needs
Fast analysis of nitroaromatics using a Thermo Scientific Hypersil GOLD PFP HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of nitroaromatics. The separation is performed on a Thermo Scientific Hypersil GOLD PFP HPLC column, with UV detection at 254 nm.
Market: Chemical
Keywords: 2,4,6-Trinitrotoluene, 2,6-Dinitrotoluene, 4-Nitrotoluene, HPLC, Hypersil Gold PFP, Nitroaromatics, Nitrotoluene, UV detection
Matrix: N/A
Uploaded on 8/30/2012.
For Research Use Only. Not for use in diagnostic procedures.