Find methods for your needs
Rapid quantification of 12 nitrosamines in metformin using triple quadrupole GC-MS/MS with Advanced Electron Ionization (AEI)
Description
The aim of this application note is to demonstrate the suitability of the Thermo Scientific™ TSQ™ 9000 triple quadrupole mass spectrometer coupled to the Thermo Scientific™ TRACE™ 1310 gas chromatograph for the analysis of 12 nitrosamines in drug substance. The linearity of the new Thermo Scientific™ TSQ™ 9610 triple quadrupole mass spectrometer NeverVent AEI equipped with the DynaMax™ XTEND XR electron multiplier detector was also tested for nitrosamines.| Market: | Pharma |
| Keywords: | Pharmaceutical, Nitrosamines, GC-MS/MS, TSQ 9610 mass spectrometer, N-nitrosodimethylamine (NDMA), N-nitrosodiethylamine (NDEA), N-nitrosodiisopropylamine (NDIPA), N-nitrosoethylisopropylamine (NEIPA) |
| Matrix: | Metformin API |
| Author: | David Lee[[1]], Aaron Lamb[[1]], Andrea Romano[[2]], and Giulia Riccardino[[2]] |
| Affiliation: | [[1]]Thermo Fisher Scientific, Runcorn, UK [[2]]Thermo Fisher Scientific, Milan, Italy |