Find methods for your needs
Uninterrupted analysis of volatile organic compounds in drinking water using single quadrupole GC-MS
Description
This application demonstrates a routine analytical method that meets the requirements outlined in U.S. EPA Method 524.2 for the quantitation of volatile organic compounds in drinking water, using the Teledyne Tekmar Atomx XYZ purge and trap (P&T) system along with a Thermo Scientific™ ISQ™ 7610 MS system coupled with a Thermo Scientific™ TRACE™ 1610 Gas Chromatograph (GC) and Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS). Method linearity, method detection limit (MDL), precision, and accuracy were assessed to evaluate method performance.| Market: | Water Analysis; Environmental; Food and Beverage |
| Keywords: | Phthalates, sensitivity, EPA, GRO, trace analysis, Gas Chromatography, selected ion monitoring, single quadrupole mass spectrometry, environmental sample analysis, THM, VOCs, environmental lab, volatiles, analytical testing laboratories |
| Matrix: | Water |
| Author: | Adam Ladak¹, Terry Jeffers², and Amy Nutter² |
| Affiliation: | ¹Thermo Fisher Scientific, USA ²Teledyne Tekmar, Mason, OH, USA |
For Research Use Only. Not for use in diagnostic procedures.