Find methods for your needs
Analysis of multiple matrices with a single calibration curve for polycyclic aromatic hydrocarbons (PAHs) with the ISQ 7610 GC-MS system following EPA Method 8270E Be the first to rate this application
analysis multiple matrices with a single calibration curve for polycyclic aromatic hydrocarbons pahs with isq 7610 gcms system following epa method 8270e
Description
The aim of this application note is to demonstrate the wide dynamic range and the robustness of the Thermo Scientific™ ISQ™ 7610 single quadrupole mass spectrometer, using the new Thermo Scientific™ XLXR™ detector, coupled to a Thermo Scientific™ TRACE™ 1610 gas chromatograph, for the analysis of 19 polycyclic aromatic hydrocarbons (PAHs) in soil and water, according to the United States Environmental Protection Agency (EPA) Method 8270E.
Market: Environmental
Keywords: Chromeleon, Polycyclic aromatic hydrocarbons (PAHs), ExtractaBrite, Environmental Analysis Pack, EPA Method 8270E, extended dynamic range, gas chromatography-mass spectrometry (GC-MS), ISQ 7610 mass spectrometer
Matrix: Soil, Water
Author: Chiara Calaprice[[1]], Brian Pike[[2]], Adam Ladak[[3]], and Paul Silcock[[3]]
Affiliation: [[1]]Thermo Fisher Scientific, Bremen, DE [[2]]PACE Analytical IDEA Laboratory, Minneapolis, MN, USA [[3]]Thermo Fisher Scientific, Hemel Hempstead, UK
Uploaded on 3/7/2022.