 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
AN73988: An Improved Method for Galactosyl Oligosaccharide Characterization
                                Description
Here, we describe a HPAE-PAD/MS method for high-resolution profiling of GOS. Separation of individual oligosaccharides in GOS was achieved on a Thermo Scientific™ Dionex™ CarboPac™ PA300-4μm column. For mass spectrometry, the oligosaccharides pass through a desalting device that replaces sodium present in the eluent for hydronium ions. Intact oligosaccharides are first analyzed without fragmentation. Then, oligosaccharide fragmentation in the negative ionization mode provides MS2 spectra that include glycosidic as well as cross-ring fragments that that were used to identify oligosaccharide DP.| Market: | Food and Beverage | 
| Keywords: | GOS, HPAE-PAD, HRAM, Q Exactive, prebiotics, TN 71, TN 20, TN 70671, TN20, TN70671, Dionex CarboPac PA300 column, desalter, Orbitrap mass analyzer, AN 73988 | 
| Matrix: | DI water | 
| Author: | Sachin Patil and Jeffrey Rohrer | 
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA, USA | 
For Research Use Only. Not for use in diagnostic procedures.