Find methods for your needs
AN000472: HPAE-PAD Method for Determination of Hib Capsular Polysaccharide Content
Description
This work describes an HPAE-PAD method for the determination of ribitol in Hib capsular polysaccharide acid hydrolysis samples. Separation of ribitol from TFA acid hydrolysis products was achieved under isocratic elution conditions using a Thermo Scientific™ Dionex™ CarboPac™ MA1 column. The method proposed here was validated for performance with respect to linearity, precision, sensitivity, and accuracy. The acid hydrolysis samples were also tested using a palladium hydrogen reference electrode.| Market: | BioPharma |
| Keywords: | Dionex CarboPac MA1 Column, Dionex ICS-5000+ System, vaccine, TN71, TN 71, Dionex ICS-6000 system, TN 20, TN 70671, TN20, TN70671, Haemophilus influenzae, ribitol, palladium hydrogen reference electrode, AN 000472, PdH |
| Matrix: | DI water |
| Author: | Sachin Patil and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA, USA |
For Research Use Only. Not for use in diagnostic procedures.