Find methods for your needs
AN000282: Determination of Glyphosate and AMPA in Oat Flour by IC-MS
Description
This study described the simultaneous direct determination of glyphosate and AMPA in oat flour by IC-MS. Glyphosate and AMPA can be determined sensitively and accurately using a Dionex IonPac AS19-4 μm column and ISQ EC single quadrupole mass spectrometer. The Reagent-Free ion chromatography system provides excellent reproducibility, thereby yielding greater quantification accuracy and consistently reliable results.| Market: | Food and Beverage |
| Keywords: | Dionex IonPac AS19-4μm column, Single quadrupole mass spectrometer, Dionex Integrion HPIC, ISQ EC, Dionex ADRS 600 suppressor, AN 000282 |
| Matrix: | Water |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.