Find methods for your needs
AN000280: Determination of Sugars in Foods using HPAE-PAD in Dual Eluent Generation Cartridge Mode
Description
In this document we demonstrate that AOAC 2018.16, a first action method for sugars determination in foods, can be executed with a Thermo Scientific™ Dionex™ CarboPac™ PA20-1 mm column using KOH/KMSA eluent produced electrolytically. using Dual Eluent Generation Cartridge (Dual EGC) mode| Market: | Food and Beverage |
| Keywords: | AOAC, Carbohydrates, Electrochemical detection, Dionex ICS-6000 HPIC system, Dual EGC Mode, TN 20, TN 70671, TN20, TN70671, AN 000280, CarboPac PA20-1 mm column |
| Matrix: | Water |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.