Find methods for your needs
A multiresidue method for pesticide profiling using an Orbitrap Tribrid mass spectrometer
Description
An AcquireX workflow implemented on the Orbitrap ID-X Tribrid mass spectrometer was evaluated for the analysis of 250 pesticides spiked into strawberry matrix to demonstrate the efficiency of the workflow for identifying the pesticides spiked at different levels. Detection and identification were determined by matching experimentally acquired MS and MS/MS data to validated spectral libraries. The efficiency of the AcquireX workflow was evaluated against a standard data-dependent acquisition and dynamic exclusion method that did not utilize an exclusion list.| Market: | Food and Beverage |
| Keywords: | Vanquish UHPLC, TraceFinder, AcquireX, Compound Discoverer, data dependent acquisition, dynamic exclusion, high-resolution accurate-mass, identification points, intelligent MS, nontargeted, Orbitrap ID-X mass spectrometer, part per billion (ppb), pesticide residues screening, quantitation of pesticide residue, Tribrid |
| Matrix: | Strawberry |
| Author: | Ed George, Seema Sharma, Scott Peterman, Richard Fussell |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.