Find methods for your needs
CAN73265: Targeted Metabolomics Method using IC Orbitrap with High Resolution Mass Spectrometry
Description
A number of different water-soluble compound standards were used, and the retention time and MS/MS spectra for more than 350 types of compounds that can be analyzed with this system were acquired to construct a metabolite library. Additionally, using cultured animal cells and human serum samples, optimization of pretreatment for the IC Q Exactive system was studied. Using the IC Q Exactive system, the conditions for IC separation were optimized, and MS and MS/MS spectral libraries were constructed.| Market: | Metabolomics |
| Keywords: | Amino acids, Nucleotides, Sugar phosphates, Organic acids, IonPac AS11-HC, metabolomics, cell extracts, CAN 73265 |
| Matrix: | Blood, Cell extracts from HeLa cells, Serum |
| Author: | Yoshihiro Izumi and Takeshi Bamba |
| Affiliation: | Kyushu University, Research Center for Transomics Medicine, Division of Metabolomics |
For Research Use Only. Not for use in diagnostic procedures.