Find methods for your needs
AN000069: Determination of Iodide in Multivitamin-Mineral Supplements using Ion Chromatography
Description
Iodine is an essential mineral for our body. This application note demonstrates the development and validation of a 15 min IC-ED method for the determination of iodide in multivitamin products. The method uses a 2 mm Dionex IonPac AG20/AS20 column set with electrolytically generated potassium hydroxide (KOH) eluent and a disposable silver working electrode. Using this method, iodide is resolved from all common anions and organic acids found in the multivitamin samples. The method has a linear calibration, is sensitive, precise, and accurate.| Market: | Food and Beverage |
| Keywords: | Dionex IonPac AS20 Column, Electrochemical detection, food, silver working electrode, RFIC, supplements, nutrition, pulsed amperometric detection (PAD), palladium hydrogen (PdH) reference electrode, Ag/AgCl pH reference, AN 000069 electrode |
| Matrix: | Food, Supplements |
| Author: | Hua Yang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.