Find methods for your needs
Fast analysis of parabens using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of parabens. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 214 nm.| Market: | Food and Beverage |
| Keywords: | Butyl paraben, Ethyl paraben, HPLC, Hypersil GOLD, Methyl paraben, Parabens, Propyl paraben, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.