Find methods for your needs
QAR Chromatography for the Dionex IonPac CS19-4µm, 2 mm Column
Description
This record has QAR conditions and results for the 2 mm Dionex IonPac CS19-4µm column. The column must be pre-conditioned offline with acetonitrile and methanesulfonic acid (MSA). See instructions in the column manual (attached). The column is optimized for small, hydrophilic amines such as ethanolamines, methylamines, ethylamines, as well as the biogenic amines.| Market: | BioPharma; Chemical; Petrochem, Oil and Gas; Food and Beverage; Environmental; Nuclear |
| Keywords: | RFIC, Alkanolamines, Biogenic Amines, IonPac CS19 column, preconditioning, alkylamines, Dionex IonPac CS19-4um column |
| Matrix: | Deionized water |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |