Find methods for your needs
Vanquish Neo UHPLC system sets new performance standards for single-shot nanoLCMS bottom-up proteomics Be the first to rate this application
vanquish neo nanolcms bottomup proteomics
Description
Demonstrate the superior performance of the Thermo Scientific™ Vanquish™ Neo, next-generation low-flow UHPLC system, for nanoLCMS bottom-up proteome profiling when coupled to a Thermo Scientific™ Orbitrap Exploris™ 480 mass spectrometer using the 75 μm I.D. × 75 cm Thermo Scientific™ EASY-Spray™ PepMap Nep column. Demonstrate system versatility and potential for new levels of proteomic depth coverage through the coupling of two 75 cm long nano columns.
Market: Proteomics
Keywords: Orbitrap Exploris 480, 1500 bar, 75 μm × 75 cm, bottom-up proteomics, deep proteome coverage, double nanoViper PepMap Neo column, EASY-Spray PepMap Neo column, HeLa protein digest, pressure, PRTC standard, Vanquish Neo UHPLC system
Matrix: HeLa protein digest
Author: R.Zheng, C.Pynn, X.Sun, B.H. Robson, A.Valenta, M.Serys, L.Taujenis, K.Woodmansey, S.Meding, W.Decrop, M.Samonig, and A.Boychenko
Affiliation: Thermo Fisher Scientific, Germering, Germany Sunnyvale, CA, USA, Vilnius, Lithuania, Hemel Hempstead, UK
Uploaded on 6/22/2021.
For Research Use Only. Not for use in diagnostic procedures.