Find methods for your needs
AN74121: Determination of Glycoprotein Sialic Acid Composition using HPAE-PAD in Dual Eluent Generation Cartridge Mode
Description
This application note demonstrates that the AU180 method for sialic acid determination of glycoproteins could be successfully modified for use with a Dionex CarboPac PA20-1 mm column and HPAE-PAD in dual eluent generation cartridge mode.| Market: | BioPharma |
| Keywords: | Electrochemical detection, Sialic acids, Dionex ICS-6000 HPIC system, Dual EGC Mode, Dionex CarboPac PA20-1 mm column, N-acetylneuraminic acid (Neu5Ac), N-glycolylneuraminic acid (Neu5Gc), AN 74121, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Acid-hydrolyzed glycoprotein |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.