Find methods for your needs
Separation of different form of plasmid DNA using anion exchnage non poros polymeric column
Description
Thermo scientific BioLC ultimate 3000RS instrument is used for the analysis of different form of plasmid DNA. The analysis is performed on high resolution anion exchange DNAPac PA200RS column.The result obtain shows well separation of linear and supercoiled form of plasmid DNA.| Market: | BioPharma |
| Keywords: | Biotherapeutics, DNA, chromatography, nucleotide, plasmid |
| Matrix: | N.A |
For Research Use Only. Not for use in diagnostic procedures.