Find methods for your needs
TN74093: Designing Ion Chromatography Methods for Determining Amines in Pharmaceuticals - Method 2
Description
This technical note (TN) describes how to develop an ion chromatography method to determine an amine in pharmaceutical samples. Two methods were developed in this TN. Method choice depends on the nature of the sample. This record is for the method that uses the Dionex IonPac CS19 column .| Market: | BioPharma |
| Keywords: | Dionex IonPac CS19 Column, Diphenhydramine, Losartan, Metformin, Ranitidine, Nitrosamines, NDMA, Dionex ICS-5000+ HPIC, Dionex CDRS 600 suppressor, Dionex ICS-6000 HPIC, TN 74093 |
| Matrix: | Pharmaceuticals |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.