Find methods for your needs
AN00010: Trace Borate Determination in High-Purity Waters
Description
In this application note, we demonstrate and evaluate an ion-exclusion method for the determination of trace-level borate in high-purity water. This ion-exclusion method coupled with suppressed conductivity detection offers an integrated solution. We used a Dionex™ IonPac™ ICE-Borate analytical column and a Dionex™ IonPac™ TBC-1 Trace Borate concentrator column, specifically designed for this application. The experimental approach shown combines large sample volume/preconcentration with ion exclusion chromatography. This approach achieves sensitive detection at low to sub-μg/L levels.| Market: | Electronics & Semiconductors |
| Keywords: | boric acid, ion-exclusion chromatography, ICE, Dionex ICS-6000 system, TN73982, Dionex IonPac ICE Borate column, Dionex ACRS-ICE 500 suppressor, TBC-1, trace borate concentrator column, AN 00010 |
| Matrix: | High-purity waters |
| Author: | Manali Aggrawal; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.