Find methods for your needs
Increase sensitivity in UHPLC-MS analysis by minimizing post-column dispersion
Description
Reduce post-column dispersion with small ID capillaries for fast gradient UHPLC-MS methods using sub-2 μm particle next-generation columns| Market: | Food and Beverage |
| Keywords: | Pesticides, UHPLC, ISQ EC, ISQ EM, Vanquish Horizon, System dispersion, high-sensitivity analysis, post-column dispersion |
| Matrix: | Methanol, Water |
| Author: | Soo Hyun Park, Mauro De Pra, Giorgia Greco |
| Affiliation: | Thermo Fisher Scientific, Germering, Germany |
For Research Use Only. Not for use in diagnostic procedures.