Find methods for your needs
Improved analysis of pesticides using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of pesticides. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 214 nm.| Market: | Environmental |
| Keywords: | A-Endosulfan, Alachlor, B-Endosulfan, Chlorfenvinphosethyl, Chloridazon, Cyanazine, Desethylatrazine, Desisopropylatrazine, Diuron, Hexazinone, HPLC, Hypersil GOLD, Linuron, Metolachor, Metoxuron, Monuron, Parathion-ethyl, Pesticides, Propazine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.