Find methods for your needs
Robust analysis of taste and odor compounds in drinking water using purge and trap and single quadrupole GC-MS
Description
To demonstrate the use of the Thermo Scientific TRACE 1310 Gas chromatograph (GC) coupled to the Thermo Scientific ISQ7000 VPI mass spectrometer (MS) and the Teledyne Tekmar Atomx XYZ purge and trap (P&T) system for the analysis of geosmin and 2-methylisoborneol (2-MIB) in drinking water according to Standard Method 6040C. Linearity, MDLs, accuracy, precision and robustness will be assessed.| Market: | Environmental |
| Keywords: | Drinking water, Gas Chromatography, selected ion monitoring, single quadrupole mass spectrometry, environmental sample analysis, purge and trap, VOCs, volatile organic compounds, analytical testing laboratories, P&T, 2-methylisoborneol, 6040, 6040C, geosmin, taste and odor |
| Matrix: | Drinking water |
| Author: | David Lee, Adam Ladak, Terry Jeffers and Amy Nutter |
| Affiliation: | Thermo Fisher Scientific, Teledyne Tekmar |
For Research Use Only. Not for use in diagnostic procedures.