Find methods for your needs
AN73981: Determination of Chlorate and Perchlorate in Milk using Ion Chromatography-Mass Spectrometry
Description
Chlorate and other oxychlorine species have been associated with inhibition of iodine uptake in humans and the formation of methemoglobin. Infants and young children are at high‐risk due to their greater dependency on milk products and higher consumption/body weight ratio. This method uses a Dionex™ IonPac™ AS20 column set, on a compact IC system (Thermo Scientific™ Dionex™ Integrion™ HPIC system) coupled with a Thermo Scientific™ ISQ EC™ single quadrupole mass spectrometer to quantify chlorate and perchlorate in milk with a simple sample preparation.| Market: | Food and Beverage |
| Keywords: | IC-MS, Integrion HPIC, ISQ EC Single Quadrupole Mass Spectrometer, Dionex IonPac AS20 column, AN 73981, Contaminant |
| Matrix: | Milk |
| Author: | Beibei Huang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.