Find methods for your needs
AN73986: Determination of Trans-Galactooligosaccharides in Foods using HPAE-PAD in Dual Eluent Generation Cartridge Mode
Description
To demonstrate that AOAC Method 2001.02 for Trans-Galactooligosaccharides (TGOS) determination in foods can be executed with a Thermo Scientific™ Dionex™ CarboPac™ PA1-1mm column using KOH/KMSA eluent produced electrolytically using Dual Eluent Generation Cartridge (Dual EGC) mode.| Market: | Food and Beverage |
| Keywords: | Electrochemical detection, GOS, TGOS, Dionex ICS-6000 HPIC system, Dionex CarboPac PA1-1mm column, Dual EGC Mode, AN 73986, Dietary Supplement, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Food |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.