Find methods for your needs
AN73901: Accurate Method for Lactose Determination in “Lactose-Free” and “Low-Lactose” Labeled Food Products
Description
This new IC method that includes sample preparation and four-micron column technology allows lactose determinations in non-dairy food samples labeled as low-lactose and lactose-free. The described instrumental setup includes a rinsing step to ensure good retention time stability in these complex samples. The method was shown to confirm “lactose-free” designations of foods sold in France that must conform to a 0.01% limit.| Market: | Food and Beverage |
| Keywords: | HPIC, HPAE-PAD, Sample preparation, Dionex ICS-6000 system, eluent generation, AN 73901, CarboPac PA210-Fast-4μm column, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Food |
| Author: | Sylvain Morel, Jeff Rohrer and Jean-François Garnier |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.