Find methods for your needs
AN73896: Carbohydrate Analysis of Agave Syrup using HPAE-PAD in Dual Eluent Generation Cartridge Mode
Description
This application demonstrates that the Norma Oficial Mexicana (Official Mexican Standard) method for agave syrup carbohydrate analysis can be executed with a Thermo Scientific™ Dionex™ CarboPac™ PA1-1mm column using KOH/KMSA eluent produced electrolytically using Dual Eluent Generation Cartridge (Dual EGC) mode.| Market: | Food and Beverage |
| Keywords: | Electrochemical detection, fructan, oligosaccharides, Adulteration, HPAE-PAD, Dionex ICS-6000 HPIC system, polyols, Mexico NOM, Dionex CarboPac PA1-1mm column, AN 73896, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Food |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.