Find methods for your needs
Analysis of underivatized amino acids in wines by solid phase extraction and HILIC-MS Be the first to rate this application
Description
Solid phase extraction cleans up wines reproducibly and reliably, facilitating analysis of underivatized amino acids in wines. Mass detection enables the quantification of co-eluting amino acids, in comparison with method involving derivatization and UV or FL detection.
Market: Food and Beverage
Keywords: HILIC, wine, ISQ EC, ISQ EM, single quadrupole mass detection, Underivatized amino acids, solid phase extraction, HyperSep C18
Matrix: Acetonitrile, Water
Author: Soo Hyun Park, Mauro De Pra, Sylvia Grosse, Stephan Meding, Frank Steiner
Affiliation: Thermo Fisher Scientific, Germering, Germany
Uploaded on 12/17/2020.
For Research Use Only. Not for use in diagnostic procedures.