Find methods for your needs
AN73854: Determination of Chlorate and Perchlorate in Powdered Infant Formula using Ion Chromatography-Mass Spectrometry
Description
Food can be contaminated by chlorate and perchlorate during different stages of production. Recent reports by the European Food Safety Authority (EFSA), found that current levels of chlorate in drinking water and in foods were too high and could negatively impact iodine uptake especially among infants and children. This method uses a Dionex™ IonPac™ AS20 column set, on a compact IC system (Thermo Scientific™ Dionex™ Integrion™ HPIC system) coupled with a Thermo Scientific™ ISQ EC™ single quadrupole mass spectrometer to determine chlorate and perchlorate in powdered infant formula.| Market: | Food and Beverage | 
| Keywords: | Dionex IonPac AS20 Column, Baby Food, IC-MS, Integrion HPIC, ISQ EC Single Quadrupole Mass Spectrometer, AN 73854 | 
| Matrix: | Powdered infant formula | 
| Author: | Beibei Huang and Jeffrey Rohrer | 
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA | 
For Research Use Only. Not for use in diagnostic procedures.