Find methods for your needs
AN44475: Determination of Iodine-Containing Species in Seaweed using IC-ICP-MS Be the first to rate this application
iodine speciation seaweed
Description
This application note describes a method by which three iodine species (MIT, DIT, and iodide) were determined in 13 min by IC-ICP-MS. The determination of iodine species consisted of a previously published extraction procedure and a newly developed IC-ICP-MS method based on anion exchange chromatography using potassium hydroxide produced by eluent generation.
Market: Food and Beverage
Keywords: iodide, speciation, IC-ICP-MS, IonPac AS20 column, iodine excess, AN 44475
Matrix: Seaweed
Author: Carl Fisher*, Daniel Kutscher*; Ana Jerse, Jens Sloth
Affiliation: Thermo Fisher Scientific*; National Food Institute, Technical University of Denmark
Uploaded on 11/30/2020.
For Research Use Only. Not for use in diagnostic procedures.