Find methods for your needs
AU73866: Determination of Trace Anions in Borated Waters by Ion Chromatography using a Large-Volume Direct Injection Be the first to rate this application
Description
This AU updates AN166 using the Dionex ICS-6000 system with Dionex EGC 500 potassium hydroxide (KOH) and a Dionex IonPac AS14 (2 × 150 mm) column. The experimental approach shown in this work combines the direct injection, electrolytically generated tetraborate eluent to analyze lithium-containing borated waters for fluoride, chloride, nitrate, phosphate, and sulfate. The large-volume direct injection technique achieves sensitive detection at low to sub-μg/L levels without using a concentrator column, sample loading pump, and additional valve.
Market: Nuclear
Keywords: Dionex ICS-5000+ System, Dionex IonPac AS14 Column, fluoride, Nitrate, Chloride, phosphate, Sulfate, Suppressed Conductivity, Dionex EGC 500 KOH, Dionex ICS-6000 system, AN166, power plants, pressurized water reactor (PWR), lithium-containing borated waters, AU 73866
Matrix: Borated water
Author: Manali Aggrawal and Jeff Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Uploaded on 11/24/2020.
For Research Use Only. Not for use in diagnostic procedures.